Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emsy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004912217.3 | XP_004912274.1 | 2 - 3 | 563 | 64 | |
XM_004912218.3 | XP_004912275.1 | 2 - 3 | 562 | 64 | ||
XM_004912219.3 | XP_004912276.1 | 2 - 3 | 563 | 64 | ||
XM_004912220.3 | XP_004912277.1 | 2 - 3 | 562 | 64 | ||
GenBank mRNA | BC135733.1 | - | 2 - 3 | 177 | 64 | |
CT027941.2 | - | 1 - 2 | 101 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |