Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122722.1 | NP_001116194.1 | 6 - 7 | 466 | 97 | |
GenBank mRNA | BC123077.1 | - | 6 - 7 | 422 | 97 | |
BC160444.1 | - | 6 - 7 | 466 | 97 | ||
CU075813.1 | - | 6 - 7 | 427 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |