Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123426.1 | NP_001116898.1 | 2 - 3 | 824 | 107 | |
GenBank mRNA | BC160486.1 | - | 2 - 3 | 824 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |