Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123464.1 | NP_001116936.1 | 1 - 2 | 126 | 84 | |
GenBank mRNA | BC161640.1 | - | 1 - 2 | 126 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |