Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123464.1 | NP_001116936.1 | 6 - 7 | 736 | 77 | |
GenBank mRNA | BC161640.1 | - | 6 - 7 | 736 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |