Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: timm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126566.1 | NP_001120038.1 | 5 - 6 | 419 | 91 | |
GenBank mRNA | BC158346.1 | - | 5 - 6 | 419 | 91 | |
CT030353.1 | - | 5 - 6 | 460 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |