Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126678.1 | NP_001120150.1 | 3 - 4 | 287 | 104 | |
GenBank mRNA | BC159145.1 | - | 3 - 4 | 287 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |