Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126814.1 | NP_001120286.1 | 5 - 6 | 673 | 83 | |
GenBank mRNA | BC160555.1 | - | 5 - 6 | 673 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |