Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126955.1 | NP_001120427.1 | 5 - 6 | 641 | 78 | |
GenBank mRNA | BC161182.1 | - | 5 - 6 | 641 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |