Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127056.1 | NP_001120528.1 | 4 - 5 | 528 | 69 | |
GenBank mRNA | CR762124.2 | - | 4 - 5 | 528 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |