Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127056.1 | NP_001120528.1 | 6 - 7 | 1209 | 95 | |
GenBank mRNA | CR762124.2 | - | 6 - 7 | 1209 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |