Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127127.1 | NP_001120599.1 | 8 - 9 | 1086 | 69 | |
GenBank mRNA | AB474592.1 | - | 7 - 8 | 992 | 69 | |
BC161634.1 | - | 8 - 9 | 1086 | 69 | ||
CT030302.1 | - | 8 - 9 | 1039 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |