Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122791.1 | NP_001116263.1 | 451 | 67 | ||
GenBank mRNA | BC121488.1 | - | 464 | 67 | ||
BC135484.1 | - | 451 | 67 | |||
BC161016.1 | - | 451 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |