Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126531.1 | NP_001120003.1 | 1 - 2 | 205 | 82 | |
XM_012956428.2 | XP_012811882.1 | 2 - 3 | 398 | 82 | ||
XM_018089976.1 | XP_017945465.1 | 2 - 3 | 241 | 82 | ||
GenBank mRNA | BC158277.1 | - | 1 - 2 | 205 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |