Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126539.1 | NP_001120011.1 | 443 | 126 | ||
GenBank mRNA | BC158308.1 | - | 443 | 126 | ||
CR761083.2 | - | 475 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |