Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126577.1 | NP_001120049.1 | 8 - 9 | 691 | 87 | |
XM_012971526.1 | XP_012826980.1 | 8 - 9 | 792 | 87 | ||
GenBank mRNA | BC158387.1 | - | 8 - 9 | 691 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |