Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126596.1 | NP_001120068.1 | 4 - 5 | 984 | 65 | |
GenBank mRNA | BC158484.1 | - | 4 - 5 | 984 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |