Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csrnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126767.1 | NP_001120239.1 | 4 - 5 | 1223 | 67 | |
GenBank mRNA | BC160429.1 | - | 4 - 5 | 1223 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |