Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126809.1 | NP_001120281.1 | 1 - 2 | 162 | 102 | |
GenBank mRNA | BC160546.1 | - | 1 - 2 | 162 | 102 | |
CU025145.1 | - | 2 - 3 | 213 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |