Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127004.1 | NP_001120476.1 | 3 - 4 | 248 | 115 | |
GenBank mRNA | BC161330.1 | - | 3 - 4 | 248 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |