Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127045.1 | NP_001120517.1 | 4 - 5 | 791 | 94 | |
GenBank mRNA | BC161418.1 | - | 4 - 5 | 791 | 94 |
Gene Symbol: LOC101730309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090738.1 | XP_017946227.1 | 2 - 3 | 324 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |