Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127045.1 | NP_001120517.1 | 6 - 7 | 1166 | 62 | |
GenBank mRNA | BC161418.1 | - | 6 - 7 | 1166 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |