Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127045.1 | NP_001120517.1 | 7 - 8 | 1266 | 103 | |
GenBank mRNA | BC161418.1 | - | 7 - 8 | 1266 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |