Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam78a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127108.1 | NP_001120580.1 | 2 - 3 | 726 | 95 | |
GenBank mRNA | BC161564.1 | - | 2 - 3 | 726 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |