Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127156.1 | NP_001120628.1 | 3 - 4 | 472 | 99 | |
XM_018095277.1 | XP_017950766.1 | 3 - 4 | 515 | 99 | ||
GenBank mRNA | BC161780.1 | - | 3 - 4 | 472 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |