Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127156.1 | NP_001120628.1 | 8 - 9 | 1518 | 94 | |
XM_018095277.1 | XP_017950766.1 | 8 - 9 | 1561 | 94 | ||
GenBank mRNA | BC161780.1 | - | 8 - 9 | 1518 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |