Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pard6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128639.1 | NP_001122111.1 | 1 - 2 | 137 | 58 | |
GenBank mRNA | BC167691.1 | - | 1 - 2 | 137 | 58 | |
CR760394.2 | - | 1 - 2 | 140 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |