Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130201.1 | NP_001123673.1 | 6 - 7 | 752 | 98 | |
GenBank mRNA | BC166964.1 | - | 6 - 7 | 752 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |