Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130350.1 | NP_001123822.1 | 11 - 12 | 1843 | 72 | |
XM_012958656.2 | XP_012814110.1 | 12 - 13 | 2001 | 72 | ||
XM_012958657.2 | XP_012814111.1 | 12 - 13 | 2028 | 72 | ||
GenBank mRNA | BC167630.1 | - | 11 - 12 | 1843 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |