Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130350.1 | NP_001123822.1 | 7 - 8 | 1488 | 70 | |
XM_012958656.2 | XP_012814110.1 | 8 - 9 | 1646 | 70 | ||
XM_012958657.2 | XP_012814111.1 | 8 - 9 | 1673 | 70 | ||
GenBank mRNA | BC167630.1 | - | 7 - 8 | 1488 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |