Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpnmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131042.1 | NP_001124514.1 | 8 - 9 | 1500 | 90 | |
GenBank mRNA | BC168052.1 | - | 8 - 9 | 1500 | 90 | |
CU075468.1 | - | 8 - 9 | 1322 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |