Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141996.1 | NP_001135468.1 | 3 - 4 | 861 | 69 | |
XM_012960677.2 | XP_012816131.1 | 3 - 4 | 1383 | 69 | ||
XM_018092943.1 | XP_017948432.1 | 2 - 3 | 254 | 69 | ||
GenBank mRNA | BC168586.1 | - | 3 - 4 | 861 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |