Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142019.1 | NP_001135491.1 | 3 - 4 | 404 | 107 | |
XM_018095294.1 | XP_017950783.1 | 2 - 3 | 414 | 107 | ||
GenBank mRNA | BC167890.1 | - | 3 - 4 | 404 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |