Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142074.1 | NP_001135546.1 | 10 - 11 | 2761 | 77 | |
XM_018092947.1 | XP_017948436.1 | 10 - 11 | 2807 | 77 | ||
GenBank mRNA | BC168023.1 | - | 10 - 11 | 2761 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |