Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c16orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142078.1 | NP_001135550.1 | 5 - 6 | 522 | 64 | |
GenBank mRNA | BC168030.1 | - | 5 - 6 | 522 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |