Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142106.1 | NP_001135578.1 | 1 - 2 | 487 | 75 | |
XM_018094569.1 | XP_017950058.1 | 1 - 2 | 522 | 75 | ||
GenBank mRNA | BC124570.1 | - | 1 - 2 | 836 | 75 | |
BC168085.1 | - | 1 - 2 | 487 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |