Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142195.1 | NP_001135667.1 | 2 - 3 | 597 | 101 | |
GenBank mRNA | BC168576.1 | - | 2 - 3 | 597 | 101 | |
CT027881.2 | - | 1 - 2 | 504 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |