Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142821.1 | NP_001136293.1 | 1 - 2 | 164 | 87 | |
GenBank mRNA | FJ428265.1 | - | 1 - 2 | 164 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |