Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142898.1 | NP_001136370.1 | 2 - 3 | 216 | 102 | |
XM_012968189.2 | XP_012823643.1 | 2 - 3 | 426 | 102 | ||
XM_012968190.2 | XP_012823644.1 | 2 - 3 | 221 | 102 | ||
GenBank mRNA | BC075483.1 | - | 2 - 3 | 270 | 102 | |
BC136184.1 | - | 2 - 3 | 222 | 102 | ||
BC161434.1 | - | 2 - 3 | 216 | 102 | ||
CT027963.1 | - | 2 - 3 | 223 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |