Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp4f22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015810.1 | NP_001015810.1 | 9 - 10 | 1271 | 113 | |
XM_012956902.2 | XP_012812356.1 | 10 - 11 | 1419 | 113 | ||
XM_012956903.2 | XP_012812357.1 | 10 - 11 | 1371 | 113 | ||
XM_012956904.2 | XP_012812358.1 | 9 - 10 | 1127 | 113 | ||
GenBank mRNA | BC090091.1 | - | 10 - 11 | 1311 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |