Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005709.1 | NP_001005709.1 | 1 | 648 | 75 | |
GenBank mRNA | BC075291.1 | - | 1 | 648 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |