Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008188.1 | NP_001008189.1 | 4 | 841 | 102 | |
GenBank mRNA | BC082514.1 | - | 4 | 841 | 102 | |
CR761594.2 | - | 4 | 839 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |