Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008447.2 | NP_001008447.2 | 4 | 1667 | 103 | |
GenBank mRNA | BC080456.1 | - | 3 | 1352 | 103 | |
CT030107.1 | - | 3 | 1663 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |