Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113159.2 | NP_001106630.1 | 1 | 452 | 113 | |
XM_012971795.2 | XP_012827249.1 | 2 | 559 | 113 | ||
GenBank mRNA | BC155515.1 | - | 1 | 453 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |