Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114645.1 | NP_001108117.1 | 1057 | 82 | ||
GenBank mRNA | AY450646.1 | - | 1034 | 82 | ||
AY773476.1 | - | 1057 | 82 | |||
BT039824.1 | - | 1218 | 82 | |||
BT053947.1 | - | 1136 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Zea mays |