Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100191272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136706.1 | NP_001130178.1 | 489 | 88 | ||
GenBank mRNA | BT033318.1 | - | 489 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |