Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cl15444_1(721) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152502.1 | NP_001145974.1 | 648 | 109 | ||
GenBank mRNA | BT054355.1 | - | 648 | 109 | ||
EU966439.1 | - | 692 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Zea mays |