Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111538.1 | NP_001105008.1 | 2116 | 76 | ||
GenBank mRNA | AF290414.1 | - | 2116 | 76 | ||
AF290415.1 | - | 1607 | 76 | |||
AY109620.1 | - | 2116 | 76 |