Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111613.1 | NP_001105083.1 | 54 | 72 | ||
GenBank mRNA | AF453838.1 | - | 54 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |