Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chb102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147851.1 | NP_001141323.1 | 273 | 76 | ||
GenBank mRNA | BT041066.1 | - | 273 | 76 | ||
EU965400.1 | - | 300 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |